Learn More
RAN Mouse anti-Human, Unconjugated, Clone: 1B11E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67500-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The RAN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAN. This antibody recognizes human, mouse, rat antigen. The RAN antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
RAN | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence | |
1B11E1 | |
Unconjugated | |
20 μL | |
5901 | |
IgG2b |