missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21107-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The RAN binding protein 10 (RANBP10) is a cytoplasmic guanine nucleotide exchange factor that shares high sequence similarity to the MET-interacting protein RANBP9. RANBP10 is expressed in the cytoplasm of mature megakaryocytes (MKs) and platelets where it localizes to polymerized noncentrosomal microtubules. RANBP10 possesses guanine nucleotide exchange factor activity towards RAN, a GTPase involved in nucleocytoplasmic traffic, spindle formation at mitosis, and nuclear envelope assembly during telophase, suggesting that RANBP10 may also be involved in these activities. RANBP10 depletion in vitro caused the disturbance of polymerized filaments in MKs and platelets in RANBP10-null mice exhibited disorders in microtubule filament numbers and localization, indicating that RANBP10 may play a significant role in maintaining microtubule dynamics.Specifications
RANBP10 | |
Polyclonal | |
Unconjugated | |
RANBP10 | |
KIAA1464, RAN binding protein 10, RANBP10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361396, 57610, 74334 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6VN19, Q6VN20 | |
Ranbp10 | |
RANBP10 Fusion Protein Ag15387 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |