missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ RanBP16 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518241
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is predicted to react with bovine, canine, mouse and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
The transport of protein and large RNAs through the nuclear pore complexes is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN plays a key role in NLS-dependent protein import. RAN-binding protein-16 is a member of the importin-beta superfamily of nuclear transport receptors.
Specifikationer
| RanBP16 | |
| Polyclonal | |
| Unconjugated | |
| Xpo7 | |
| 4930506C02Rik; BB164534; EXP7; exportin 7; exportin-7; Kiaa0745; mKIAA0745; RAN binding protein 16; ran-binding protein 16; Ranbp16; Xpo7 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23039 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UIA9 | |
| Xpo7 | |
| Synthetic peptide sequence (NSTYGVNSNDMMS) corresponding to the C-terminus amino acids of XPO7 (aa 1075-1087). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering