missing translation for 'onlineSavingsMsg'
Learn More

RANBP3L Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17875-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16837874

  • 1656.73 SEK / 20µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RANBP3L
Polyclonal
Unconjugated
RANBP3L
RAN binding protein 3 like, RANBP3L
Rabbit
Antigen Affinity Chromatography
RUO
202151
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q86VV4
RANBP3L
RANBP3L Fusion Protein Ag11642
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers