missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP3L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17875-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
RANBP3L | |
Polyclonal | |
Unconjugated | |
RANBP3L | |
RAN binding protein 3 like, RANBP3L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
202151 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q86VV4 | |
RANBP3L | |
RANBP3L Fusion Protein Ag11642 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |