Läs mer
Ranimustine, TRC
Produktkod. 30415717
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30415717 25 mg
1 options
Produktkod. 30415717 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR12003025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ranimustine
CAS 58994-96-0
Molekylformel C10 H18 Cl N3 O7
Kvantitet 25 mg
Synonym α-D-Glucopyranoside','methyl 6-[[[(2-chloroethyl)nitrosoamino]carbonyl]amino]-6-deoxy-, Methyl 6-[[[(2-chloroethyl)nitrosoamino]carbonyl]amino]-6-deoxy-α-D-glucopyranoside, Cymerin, Cymerine, MCNU, NSC 270516, Ranimustine
InChI formel InChI=1S/C10H18ClN3O7/c1-20-9-8(17)7(16)6(15)5(21-9)4-12-10(18)14(13-19)3-2-11/h5-9,15-17H,2-4H2,1H3,(H,12,18)/t5-,6-,7+,8-,9+/m1/s1
LEDER CO[C@H]1O[C@H](CNC(=O)N(CCCl)N=O)[C@@H](O)[C@H](O)[C@H]1O
IUPAC-namn 1-(2-chloroethyl)-1-nitroso-3-[[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-methoxyoxan-2-yl]methyl]urea
Molekylvikt (g/mol) 327.72
Formel vikt 327.0833