Läs mer
Ranitidine S-Oxide, TRC
Produktkod. 30008170
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008170 100 mg
30005303 50 mg
17952155 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008170 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR120015100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73851-70-4
Molekylformel C13 H22 N4 O4 S
Molekylvikt (g/mol) 330.4
Synonym N-[2-[[[5-[(Dimethylamino)methyl]furan-2-yl]methyl]sulphinyl]ethyl]-N'-methyl-2-nitroethene-1,1-diamine, Ranitidine Hydrochloride Imp. C (EP), Ranitidine Sulphoxide
IUPAC-namn 1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfinyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine
LEDER CNC(=C[N+](=O)[O-])NCCS(=O)Cc1oc(CN(C)C)cc1
Specifikationer
Kemiskt namn eller material Ranitidine Sulfoxide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C13H22N4O4S/c1-14-13(9-17(18)19)15-6-7-22(20)10-12-5-4-11(21-12)8-16(2)3/h4-5,9,14-15H,6-8,10H2,1-3H3
Formel vikt 330.1362
Procent renhet >95%