Läs mer
Ranolazine, TRC
Produktkod. 30134767
Change view
Kvantitet:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134767 100 mg
30138686 500 mg
30007437 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134767 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR122500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 95635-55-5
Molekylformel C24 H33 N3 O4
Molekylvikt (g/mol) 427.54
Synonym N-(2,6-Dimethylphenyl)-4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]-1-piperazineacetamide, 1-Piperazineacetamide, N-(2,6-dimethylphenyl)-4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]-, (±)-, (±)-Ranolazine, CVT 303, RS 43285-003, Ranexa, Ranolazine, Renexa
IUPAC-namn N-(2,6-dimethylphenyl)-2-[4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-yl]acetamide
LEDER COc1ccccc1OCC(O)CN2CCN(CC(=O)Nc3c(C)cccc3C)CC2
Specifikationer
Kemiskt namn eller material Ranolazine
Kvantitet 100 mg
InChI formel InChI=1S/C24H33N3O4/c1-18-7-6-8-19(2)24(18)25-23(29)16-27-13-11-26(12-14-27)15-20(28)17-31-22-10-5-4-9-21(22)30-3/h4-10,20,28H,11-17H2,1-3H3,(H,25,29)
Formel vikt 427.2471