Läs mer
Ranolazine, TRC
Produktkod. 30138686
Change view
Kvantitet:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138686 500 mg
30134767 100 mg
30007437 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138686 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR122500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 95635-55-5
Molekylformel C24 H33 N3 O4
Molekylvikt (g/mol) 427.54
Synonym N-(2,6-Dimethylphenyl)-4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]-1-piperazineacetamide, 1-Piperazineacetamide, N-(2,6-dimethylphenyl)-4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]-, (±)-, (±)-Ranolazine, CVT 303, RS 43285-003, Ranexa, Ranolazine, Renexa
IUPAC-namn N-(2,6-dimethylphenyl)-2-[4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-yl]acetamide
LEDER COc1ccccc1OCC(O)CN2CCN(CC(=O)Nc3c(C)cccc3C)CC2
Specifikationer
Kemiskt namn eller material Ranolazine
Kvantitet 500 mg
InChI formel InChI=1S/C24H33N3O4/c1-18-7-6-8-19(2)24(18)25-23(29)16-27-13-11-26(12-14-27)15-20(28)17-31-22-10-5-4-9-21(22)30-3/h4-10,20,28H,11-17H2,1-3H3,(H,25,29)
Formel vikt 427.2471