missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RAP2A |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892174
|
Proteintech
13789-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16882174
|
Proteintech
13789-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RAP2A is a signaling protein involved in small GTPase mediated signal transduction activated by guanine nucleotide-exchange factors (GEF) EPAC and EPAC2, in a cAMP-dependent manner, and GFR. This membrane protein belongs to the small GTPase superfamily and Ras family and contains domains required for GTP binding and a Ras-like C-terminal motif for post-translational lipid binding and subsequent anchoring to the plasma membrane. Apart from having a possible role in membrane shedding, RAP2A also functions as a slowly responding molecular switch in the RAP1A signaling cascade and also interacts with ARHGAP29, the putative specific effector of RAP2A.Specifications
RAP2A | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P10114, Q80ZJ1 | |
5911, 76108 | |
RAP2A Fusion Protein Ag4808 | |
Primary | |
-20°C | |
RAP2A |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
K REV, KREV, RAP2, RAP2A, Ras related protein Rap 2a, RbBP 30 | |
RAP2A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |