Läs mer
Rapeseed Oil, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Rapeseed Oil |
| CAS | 8002-13-9 |
| Kvantitet | 5 mL |
| Synonym | AP 85, Agri-Pure 60, Beloukha, Brassica napus oil, Brassica napus oleifera biennis seed oil, Brassica napus seed oil, Codacide, Codacide Oil, Cole seed oil, Colleseed oil, Coltza oil, Colza oil, Cutinol V 7, Ekol, Fats and Glyceridic oils','colza, Fats and Glyceridic oils','rape, Fry Ace PO, Oils','colza, Oils','glyceridic','colza, Oils','glyceridic','rape, Plantocorit N, Plantocut 10S, Plantohyd, Plantohyd 40N, Raisio Biosave 32L |