Learn More
Rapidosept, TRC
Product Code. 30427924
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
30427924 10 g 10g
1 options
Product Code. 30427924 Supplier Toronto Research Chemicals Supplier No. TRCR12450010G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-dichlorobenzyl alcohol
CAS 1777-82-8
Molecular Formula C7 H6 Cl2 O
Quantity 10 g
Synonym (2,4-Dichlorophenyl)methanol, 2,4-Dichlorobenzenemethanol, 2,4-Dichlorobenzyl alcohol, Dybenal, Myacide, Myacide SP, NSC 15635, Rapidosept
InChI Formula InChI=1S/C7H6Cl2O/c8-6-2-1-5(4-10)7(9)3-6/h1-3,10H,4H2
SMILES OCc1ccc(Cl)cc1Cl
IUPAC Name (2,4-dichlorophenyl)methanol
Molecular Weight (g/mol) 177.03
Formula Weight 175.9796