Läs mer
Rasagiline-13C3 Mesylate, TRC
Produktkod. 30132450
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132450 1 mg
30011050 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132450 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1260031MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391052-18-8
Molekylformel 13C3 C9 H13 N . C H4 O3 S
Molekylvikt (g/mol) 270.32
Synonym (1R)-2,3-Dihydro-N-2-propyn-1-yl-1H-inden-1-amine-13C3 Mesylate, (R)-2,3-Dihydro-N-2-Propynyl-1-indenamine-13C3 Mesylate, Agilect-13C3, Azilect-13C3 Mesylate, TVP-1012-13C3 Mesylate
IUPAC-namn methanesulfonic acid;(1R)-N-(1,2,3-^{13}C_{3})prop-2-ynylindan-1-amine
LEDER CS(=O)(=O)O.[13CH]#[13C][13CH2]N[C@@H]1CCc2ccccc12
Specifikationer
Kemiskt namn eller material Rasagiline-13C3 Mesylate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C12H13N.CH4O3S/c1-2-9-13-12-8-7-10-5-3-4-6-11(10)12;1-5(2,3)4/h1,3-6,12-13H,7-9H2;1H3,(H,2,3,4)/t12-;/m1./s1/i1+1,2+1,9+1;
Formel vikt 270.103
Procent renhet >95%