Learn More
RASGEF1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17121-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Guanine nucleotide exchange factor (GEF) with specificity for RAP2A, KRAS, HRAS, and NRAS (in vitro). Plays a role in cell migration.Specifications
RASGEF1A | |
Polyclonal | |
Unconjugated | |
RASGEF1A | |
CG4853, RASGEF1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221002, 312664, 70727 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N9B8 | |
Rasgef1a | |
RASGEF1A Fusion Protein Ag10809 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |