Läs mer
Rat anti-Mouse IgG2b Heavy Chain Secondary Antibody (LO-MG2b-2), DyLight 550, R&D Systems™
Produktkod. 30232453 Handla allt Bio Techne Produkter
Change view
Kvantitet:
0.25 mL
Produktkod. Kvantitet
30232453 0.25 mL
1 options
Produktkod. 30232453 Leverantör Novus Biologicals Leverantörsnummer NBP268438R
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A Novus product, part of R&D Systems. Rat Monoclonal Antibody
IgG2b Heavy Chain Monoclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen IgG2b Heavy Chain
Användningsområden ELISA
Klassificering Monoclonal
Klona LO-MG2b-2
Konjugera DyLight 550
Formulering 50mM Sodium Borate
Värd art Rat
Immunogen Purified mouse IgG2b
Reningsmetod Protein A purified
Kvantitet 0.25 mL
Regulatorisk status RUO
Primär eller sekundär Secondary
Målarter Mouse
Innehåll och lagring Store at 4°C in the dark.
Produkttyp Antibody
Form Purified
Isotyp IgG1
Produktlinje Novus