missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat anti-Mouse IgM Heavy Chain Secondary Antibody (eB121-15F9), DyLight 755, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgM Heavy Chain Monoclonal antibody specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | IgM Heavy Chain |
| Användningsområden | Flow Cytometry |
| Klassificering | Monoclonal |
| Klona | eB121-15F9 |
| Konjugera | DyLight 755 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain |
| Värd art | Rat |
| Immunogen | Mouse IgM |
| Reningsmetod | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?