Learn More
Novus Biologicals™ Rat Caveolin-1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marke:  Novus Biologicals™ NBP2-75052
Artikelnummer. 18323221
  • 7771.94 SEK / 1 set
Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
The Rat Caveolin-1 ELISA Kit (Colorimetric) measures rat Caveolin-1 in serum, plasma and other biological fluids.
Spezifikation
Spezifikation
0.16 - 10 ng/mL
This kit recognizes Rat Cav-1 in samples. No significant cross-reactivity or interference between Rat Cav-1 and analogues was observed.
Biological Fluids, Plasma, Serum
857
Rat
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
CAV1
1 Kit
Caveolin-1
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote