Learn More
Novus Biologicals™ Rat Caveolin-1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marke: Novus Biologicals™ NBP2-75052
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Rat Caveolin-1 ELISA Kit (Colorimetric) measures rat Caveolin-1 in serum, plasma and other biological fluids.Spezifikation
0.16 - 10 ng/mL | |
This kit recognizes Rat Cav-1 in samples. No significant cross-reactivity or interference between Rat Cav-1 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
857 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
0.10 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CAV1 | |
1 Kit | |
Caveolin-1 |