Learn More
Novus Biologicals™ Rat CENPE ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75230
This item is not returnable.
View return policy
Description
The Rat CENPE ELISA Kit (Colorimetric) measures rat CENPE in serum, plasma and other biological fluids.Specifications
0.16 - 10 ng/mL | |
This kit recognizes Rat CENPE in samples. No significant cross-reactivity or interference between Rat CENPE and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1062 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
0.10 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CENPE | |
1 Kit | |
CENPE |