missing translation for 'onlineSavingsMsg'
Läs mer

R&D Systems™ Rat CNTFR alpha (NP_001003929) VersaClone cDNA

Produktkod. 18719847
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18719847 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18719847 Leverantör R&D Systems™ Leverantörsnummer RDC2749
  • ONLINE EXCLUSIVE PRICE
    3475.00 SEK
    3285.45 SEK / 10µg
    Save 189.55 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 14-10-2026
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • CNTFR is a member of the type 1 cytokine receptor family.
  • CNTFR is the ligand-specific component of a tripartite receptor for ciliary neurotrophic factor, which plays a critical role in neuronal cell survival, differentiation and gene expression.
  • Binding of ciliary neurotrophic factor to CNTFR recruits the transmembrane components of the receptor, gp130 and leukemia inhibitory factor receptor, facilitating signal transduction.
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifikationer

Gen rCntfr
Tillträdesnummer NP_001003929
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl, pH 8.5
Infoga typ 1132 bp
Gen-ID (Entrez) 1271
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Art Rat
Kvantitet 10 μg
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.