Learn More
Rat IgG1 kappa Isotype Control (eBRG1), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Isotype Control
Marke: Invitrogen 64-4301-80
Weitere Details : Weight : 0.01000kg
Beschreibung
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control has been reported for use in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA. Applications Tested: Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin.Applications Reported: This rat IgG1 isotype control has been reported for use in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA.Applications Tested: Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody.Excitation: 405nm; Emission: 645nm; Laser: Violet LaserSpezifikation
Rat IgG1 kappa | |
eBRG1 | |
Super Bright 645 | |
Rat | |
25 μg | |
Isotype Control (Primary) | |
4°C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
Flow Cytometry | |
0.2mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Affinity chromatography | |
RUO | |
None | |
Liquid |