missing translation for 'onlineSavingsMsg'
Läs mer

Rat IgG2a kappa Isotype Control (eBR2a), Functional Grade, eBioscience™, Invitrogen™

Produktkod. 17181232
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17181232 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17181232 Leverantör Affymetrix eBioscience Leverantörsnummer 164321050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Rat Isotype Control

Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG2a isotype control has been reported for use in flow cytometric analysis and ELISA. Applications Tested: This rat IgG2a isotype control has been used by flow cytometric analysis of mouse splenocyte suspension and can be used at the same concentration as the experimental antibody. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

IgG2a kappa Rat, Isotype Control, Functional Grade, eBioscience™ Invitrogen™ is validated for Control, Flow Cytometry, Functional Assay.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Rat IgG2a kappa
Användningsområden Flow Cytometry, Functional Assay
Klona eBR2a
Koncentration 1 mg/mL
Konjugera Functional Grade
Formulering PBS with no preservative; pH 7.2
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Värd art Rat
Reningsmetod Affinity chromatography
Kvantitet 50 mg
Regulatorisk status RUO
Primär eller sekundär Isotype Control (Primary)
Målarter None
Innehåll och lagring 4° C
Form Liquid
Isotyp IgG2a κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.