Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), Super Bright™ 702, eBioscience™, Invitrogen™
Rat Isotype Control
1638.00 SEK - 3885.00 SEK
Specifications
Antigen | Rat IgG2b kappa |
---|---|
Clone | eB149/10H5 |
Conjugate | Super Bright 702 |
Form | Liquid |
Host Species | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15895733
|
Affymetrix eBioscience
67-4031-82 |
100 μg |
3885.00 SEK
100µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15885733
|
Affymetrix eBioscience
67-4031-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2b kappa | |
Super Bright 702 | |
Rat | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Isotype Control (Primary) |
eB149/10H5 | |
Liquid | |
RUO | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only