missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Gibco™ Rat IL-5 Recombinant Protein, PeproTech®
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
100 μg
2 μg
2.5 x 200 μg
200 μg
5 x 200 μg
50 μg
Beskrivning
400-05-1MG will be provided as 5 x 200 μg (400-05-200UG). 400-05-500UG will be provided as multiple 100 μg (400-05-100UG) and 200 μg (400-05-200UG) vials. Recombinant Rat IL-5 is a 26.0 kDa disulfide-linked homodimeric protein containing two 113 amino acid chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
Specifikationer
Specifikationer
| Tillträdesnummer | Q08125 |
| För användning med (applikation) | Functional Assay |
| Formulering | protein with no preservative |
| Gen-ID (Entrez) | 24497 |
| Molekylvikt (g/mol) | 26 |
| Namn | Rat IL-5 |
| Kvantitet | 2.5 x 200 μg |
| Källa | E. coli |
| Förvaringskrav | -20°C |
| Regulatorisk status | RUO |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering