missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Rat IL-2 Recombinant Protein

Produktkod. 11848811
Klicka för att se tillgängliga alternativ
Kvantitet:
20 μg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11848811

Brand: Invitrogen™ RRIL2I

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Mature rat IL-2 was expressed in Pichia pastoris, X-33. GenBank Accession # P04351, amino acids 21 - 155. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: APTSSPAKET. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant rat IL-2 was determined by measuring the proliferation of mouse CTLL-2 cells. The ED50 (or 1 unit of activity) for this effect is typically 3 ng/mL. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer P17108
För användning med (applikation) Bioactivity
Formulering PBS with no preservative
Gen-ID (Entrez) 116562
Molekylvikt (g/mol) 15.5 kDa
Namn Rat IL-2
Reningsmetod Purified
Kvantitet 20 μg
Källa Yeast
Förvaringskrav 4°C
Regulatorisk status RUO
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Vanligt namn IL-2
Gensymbol IL2
Biologisk aktivitet ED50 = 3 ng/mL; determined by the dose-dependent proliferation of mouse CTLL-2 cells.
Produkttyp Protein
Konjugera Unconjugated
Art Yeast
Rekombinant Recombinant
Sekvens Rat IL-2, amino acids 21-155 (Accession # P04351)
Innehåll och lagring 4°C
Aktivitet ED50 = 3 ng/mL; determined by the dose-dependent proliferation of mouse CTLL-2 cells.
Uttryckssystem P. pastoris
Form Lyophilized
Protein subtyp Recombinant
Renhet eller kvalitetsklass >95% by SDS-PAGE
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.