Läs mer
Rat μ Opioid R/OPRM1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8629
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Immunohistochemistry.
Specifikationer
| mu Opioid R/OPRM1 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 4988 | |
| Rat | |
| Supernatant |
| Immunohistochemistry | |
| 1126B | |
| Immunohistochemistry 1:125 dilution | |
| P33535 | |
| OPRM1 | |
| Synthetic peptide with a short amino acid sequence from the N-terminus of rat μ Opioid R/OPRM1 Accession # P33535 | |
| 100 μg | |
| Primary | |
| Detects rat μ Opioid R/OPRM1 in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |