Läs mer
R&D Systems™ Rat PDGF R beta (NP_113713.1) VersaClone cDNA
Specifikationer
Specifikationer
| Gen | PDGF R beta |
| Tillträdesnummer | NP_113713.1 |
| Koncentration | 10 μg at 0.2 μg/μL |
| Formulering | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Infoga typ | 3307 bp |
| Gen-ID (Entrez) | 5159 |
| Gene Alias | beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Art | Rat |
| Produktlinje | R&D Systems |
| Visa mer |