Learn More
Invitrogen™ Rat Prolactin ELISA Kit
Rat Prolactin quantitates rat Prolactin in serum, plasma, supernatant.
Brand: Invitrogen™ ERA50RB
3920.19 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.30000kg
Description
Rat Prolactin quantitates rat Prolactin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Prolactin.
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.Specifications
AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
0.41 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
PRL | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.41 to 100 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P01237 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL | |
Prolactin | |
4 hr 45 min |