missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat Prolactin ELISA Kit
Rat Prolactin quantitates rat Prolactin in serum, plasma, supernatant.
5880.00 SEK - 40370.00 SEK
Specifications
Accession Number | AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL |
---|---|
Assay Range | 0.41 to 100 ng/mL |
Assay Sensitivity | 0.41 ng/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17155889
|
Invitrogen™
ERA50RBX5 |
5 x 96 Tests |
28010.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
16824420
|
Invitrogen™
ERA50RB |
96 Tests |
5880.00 SEK
96 tests |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
18597894
|
Invitrogen™
ERA50RBX10 |
10 x 96 Tests |
40370.00 SEK
Pack of 10 |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rat Prolactin quantitates rat Prolactin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Prolactin.
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.Specifications
AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
0.41 ng/mL | |
ELISA | |
P01237 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
Prolactin | |
4 hr 45 min |
0.41 to 100 ng/mL | |
Biotin | |
Colorimetric Microplate Reader | |
PRL | |
<10% | |
HRP | |
2°C to 8°C | |
1 hr 20 min |