missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Rat TNF-alpha Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 27 publications
Brand: R&D Systems AF-510-NA
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TNF-alpha Polyclonal specifically detects TNF-alpha in Rat samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifikationer
| TNF-alpha | |
| Polyclonal | |
| Unconjugated | |
| P16599 | |
| TNF | |
| E. coli-derived recombinant rat TNF-alpha Leu80-Leu235 Accession # P16599 | |
| 100 μg | |
| Primary | |
| Detects rat TNF-alpha in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant porcine TNF-alpha is observed and less than 1% cross-reactivity with recombinant human (rh) TNF-beta , rhTNF sRI, and rhTNF sRII is observed. In direct ELISAs, approximately 40% cross-reactivity with recombinant canine TNF-alpha , recombinant feline TNF-alpha , and recombinant rhesus macaque TNF-alpha is observed. Neutralizes the biological activity of rat TNF-alpha . This antibody will also neutralize the biological activity of murine TNF-alpha , but weak neutralization of rhTNF-alpha and no neutralization of rhTNF-beta was observed. | |
| Rat | |
| IgG |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.15-0.75 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7124 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering