missing translation for 'onlineSavingsMsg'
Learn More

Rb Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17218-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877064

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human RB1.

ZNF639 binds DNA and may function as a transcriptional repressor.
Specifications

Specifications

Rb
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06400
OSRC, p105 Rb, pp110, pRb, Rb, RB1, retinoblastoma 1
Rabbit
IgG
150 μL
-20°C
Primary
5925
Western Blot
Unconjugated
RB1
Liquid
RB1
Rb Fusion Protein Ag11035
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers