missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1CC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | RB1CC1 |
---|---|
Concentration | 0.62 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826193
|
Proteintech
10043-2-AP-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816193
|
Proteintech
10043-2-AP-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The immunogen of this antibody is C-terminal 154aa of RB1CC1.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
RB1CC1 | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q8TDY2, Q9ESK9 | |
12421, 312927, 9821 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.62 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RB1CC1 | |
RB1CC1 | |
Primary | |
-20°C | |
RB1CC1 |