missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM12B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RBM12B |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836974
|
Proteintech
17137-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826974
|
Proteintech
17137-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RBM12B gene ontology annotations related to this gene include RNA binding.Specifications
RBM12B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80YR9, Q8IXT5 | |
313069, 389677, 72397 | |
RBM12B Fusion Protein Ag10894 | |
Primary | |
-20°C | |
RBM12B |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RBM12B, RNA binding motif protein 12B, RNA binding protein 12B | |
Rbm12b, Rbm12b1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |