missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM15 Mouse anti-Human, Clone: 4A1A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66059-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
May function as an mRNA export factor, stimulating export and expression of RTE-containing mRNAs which are present in many retrotransposons that require to be exported prior to splicing. High affinity binding of pre-mRNA to RBM15 may allow targeting of the mRNP to the export helicase DBP5 in a manner that is independent of splicing-mediated NXF1 deposition, resulting in export prior to splicing. May be implicated in HOX gene regulation.Specifications
RBM15 | |
Monoclonal | |
0.99 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96T37 | |
RBM15 | |
RBM15 Fusion Protein Ag17970 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
4A1A4 | |
Unconjugated | |
RBM15 | |
One twenty two protein 1, OTT, OTT1, RBM15, RNA binding motif protein 15, SPEN | |
Mouse | |
Protein A | |
RUO | |
64783 | |
-20°C | |
Liquid |