missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM15B Mouse anti-Human, Unconjugated, Clone: 1C2C11, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RBM15B |
---|---|
Clone | 1C2C11 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840176
|
Proteintech
67506-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830176
|
Proteintech
67506-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The RBM15B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBM15B. This antibody recognizes human antigen. The RBM15B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RBM15B | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
29890 | |
Primary |
1C2C11 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |