Learn More
RBM24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18178-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
RBM24 is preferentially expressed in muscle during differentiation in vitro and may regulate myogenic differentiation.Specifications
RBM24 | |
Polyclonal | |
Unconjugated | |
RBM24 | |
RBM24, RNA binding motif protein 24, RNA binding protein 24, RNPC6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221662, 666794, 690139 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D3Z4I3, Q9BX46 | |
Rbm24 | |
RBM24 Fusion Protein Ag12871 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |