missing translation for 'onlineSavingsMsg'
Learn More

RBP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11774-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16829073

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells.
Specifications

Specifications

RBP4
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P02753, P04916, Q00724
Plasma retinol binding protein, PRBP, RBP, RBP4, Retinol binding protein 4
Rabbit
IgG
150 μL
-20°C
Primary
19662, 25703, 5950
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RBP4
Liquid
RBP4
RBP4 Fusion Protein Ag2448
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers