missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14895-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.Specifications
RBX1 | |
Polyclonal | |
Unconjugated | |
RBX1 | |
Protein ZYP, RBX1, Regulator of cullins 1, ring box 1, RING box protein 1, RING finger protein 75, RNF75, ROC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
300084, 56438, 9978 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62877, P62878 | |
Rbx1 | |
RBX1 Fusion Protein Ag6685 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |