Learn More
Hamilton™ RCX-30 7 μm 4.1 x 250 mm
Hamilton RCX-30 columns are designed with high exchange capacity for quality carbohydrate separation. Isocratic or gradient separations can be used depending on the complexity of the mixture.
Brand: Hamilton™ 79803
Additional Details : Weight : 0.20100kg
Description
Specification Sheet
Separation Mode | Anion Exchange |
---|---|
Stationary Phase | RCX-30 |
Format | HPLC Column |
Hardware Inner Diameter | 4.1 mm |
Hardware Length | 250 mm |
Particle Size | 7 μm |
USP Classification | USP L47 |
Packing Material Type | PSDVB/Trimethylammonium |
Pore Size | 100 Å |
Hardware Material | Stainless Steel |
Max Pressure | 5,000 psi |
Exchange Capacity | 1.0 meq/gm |
Exchange Capacity/Column | 1.6 meq/column |
Mobile Phase Limits | pH 1-13. 0-100% aqueous, organic modifier |
Buffer Strength | 0.0-1.0N |
Temp Limits | pH 1-7.9 (5-60°C); pH 8-13 (5-30°C) |
Restoration | Flush with 150 mL 0.1 N sodium hydroxide. |
Compound Classification | Complex Carbohydrates |
UOM | 1 EACH |
Specifications
HPLC Column | |
250 mm | |
7 μm | |
100 Å | |
USP L47 | |
PSDVB/Trimethylammonium | |
Stainless Steel |
4.1 mm | |
5,000 psi | |
pH 1-13. 0-100% aqueous, organic modifier | |
pH 1°C to 7.9°C (5°C to +60°C); pH 8°C to 13°C (5°C to +30°C) | |
RCX-30 | |
Anion Exchange | |
High Exchange Capacity Anion Exchange Column |