missing translation for 'onlineSavingsMsg'
Learn More

RDH14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16283-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825874

  • 3313.85 SEK / 150µL
Estimated Shipment: 08-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Exhibits an oxidoreductive catalytic activity towards retinoids. Most efficient as an NADPH-dependent retinal reductase. Displays high activity toward 9-cis and all-trans-retinol. No steroid dehydrogenase activity detected.
Specifications

Specifications

RDH14
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9ERI6, Q9HBH5
Alcohol dehydrogenase PAN2, PAN2, RDH14, Retinol dehydrogenase 14, SDR7C4
Rabbit
IgG
150 μL
-20°C
Primary
105014, 500629, 57665
Western Blot
Unconjugated
RDH14
Liquid
Rdh14
RDH14 Fusion Protein Ag9386
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers