missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ REA Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA521417

Produktkod. 13238968

  • 4615.00 SEK / 100µL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

Recommended positive controls: H1299, HeLa, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (87%), Pig (98%), Chicken (95%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PHB2 acts as a mediator of transcriptional repression by nuclear hormone receptors via recruitment of histone deacetylases. This protein functions as an estrogen receptor (ER)-selective coregulator that potentiates the inhibitory activities of antiestrogens and represses the activity of estrogens. It competes with NCOA1 for modulation of ER transcriptional activity. It is probably involved in regulating mitochondrial respiration activity and in aging.
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

REA
Polyclonal
Unconjugated
PHB2
AU044498; BAP; Bap37; Bap-37; Bcap27; BCAP37; B-cell associated protein; B-cell receptor associated protein 37; B-cell receptor-associated protein 37; B-cell receptor-associated protein BAP37; D-prohibitin; hBAP; MGC117268; p22; phb2; phb2 {ECO:0000250; PNAS-141; prohibitin 2; prohibitin-2; Rea; Repressor of estrogen receptor activity; UniProtKB:Q99623}
Rabbit
Antigen affinity chromatography
RUO
11331, 114766, 12034
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.66 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O35129, Q5XIH7, Q99623
PHB2
Recombinant protein encompassing a sequence within the center region of human Prohibitin 2. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.