Learn More
Lenz Laborglasinstrumente™ Reaction vessel cylindrical round bottom with withdrawal valve 100mm flange 3L Lenz
Product Code. 11542214
Change view
Click to view available options
Capacity (Metric):
3 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
11542214 3 L Each
1 options
Product Code. 11542214 Supplier Lenz Laborglasinstrumente™ Supplier No. 06007276
  • ONLINE EXCLUSIVE PRICE
    9345.00 SEK
    7757.69 SEK / Paket med 1
    Save 1587.31 SEK
    16% Av
Beräknad leverans: 12-01-2027