Läs mer
Rebamipide, MedChemExpress
Produktkod. 16452149
Encompass_Preferred
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16452149 1 g 1g
16410898 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16452149 Leverantör Medchem Express Leverantörsnummer HYB0360
MedChemExpress Rebamipide (OPC12759) is a mucoprotective agent. Rebamipide induces COX-2 expression, increases PGE2 levels, and enhances gastric mucosal defense in a COX-2-dependent manner.
Rebamipide (OPC12759) is a mucoprotective agent. Rebamipide induces COX-2 expression, increases PGE2 levels, and enhances gastric mucosal defense in a COX-2-dependent manner.
Specifikationer
Kemiskt namn eller material Rebamipide
Fysisk form Solid
CAS 90098-04-7
Färg White
Kvantitet 1 g
Anteckningar om renhetsgrad Research
Molekylformel C19H15ClN2O4
För användning med (applikation) Cancer-programmed cell death
Synonym OPC12759 Proamipide
Löslighetsinformation DMSO : 50 mg/mL (134.85 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
LEDER OC(C(NC(C1=CC=C(Cl)C=C1)=O)CC(C2=CC=CC=C2N3)=CC3=O)=O
Molekylvikt (g/mol) 370.79
Formel vikt 370.79
Procent renhet 99.88%