Läs mer
Rebamipide, TRC
Produktkod. 30007147
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007147 5 g
30005499 1 g
17999554 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1390005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90098-04-7
Molekylformel C19H15ClN2O4
Molekylvikt (g/mol) 370.79
Synonym 4-Quinolinepropanoic acid, α-[(4-chlorobenzoyl)amino]-1,2-dihydro-2-oxo-, α-[(4-Chlorobenzoyl)amino]-1,2-dihydro-2-oxo-4-quinolinepropanoic acid, Mucosta, OPC 12759, Proamipide, Rebamipide, Rebator
IUPAC-namn 2-[(4-chlorobenzoyl)amino]-3-(2-oxo-1H-quinolin-4-yl)propanoic acid
LEDER OC(=O)C(CC1=CC(=O)Nc2ccccc12)NC(=O)c3ccc(Cl)cc3
Specifikationer
Kemiskt namn eller material Rebamipide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 g
InChI formel InChI=1S/C19H15ClN2O4/c20-13-7-5-11(6-8-13)18(24)22-16(19(25)26)9-12-10-17(23)21-15-4-2-1-3-14(12)15/h1-8,10,16H,9H2,(H,21,23)(H,22,24)(H,25,26)
Formel vikt 370.072
Procent renhet >95%