Läs mer
Rebaudioside A, TRC
Produktkod. 30134526
Change view
Kvantitet:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134526 100 g
30009355 10 g
17938894 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR139500100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58543-16-1
Molekylformel C44 H70 O23
Molekylvikt (g/mol) 967.01
Synonym Kaur-16-en-18-oic acid, 13-[(O-β-D-glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, β-D-glucopyranosyl ester, (4α)-, 4G-S, Chrysanta AR-P, Glycoside A3, from Stevia rebaudiana, Glycoside X, Pure Via, RA 95, Reb-A 97, Rebaudiana a, Rebaudioside A, Rebiana, SG 95RA50, SG 95RA60, SG 95RA70, SG 95RA80, SG 95RA85, SG 95RA90, Sooolite!-Pure, Stevioside a3, Sweetener 4G-S
LEDER C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)[C@](CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@H]5O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)C(=O)O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O
Specifikationer
Kemiskt namn eller material Rebaudioside A
Anteckningar om renhetsgrad HPLC
Kvantitet 100 g
InChI formel InChI=1S/C44H70O23/c1-17-11-43-9-5-22-41(2,7-4-8-42(22,3)40(59)66-38-33(58)30(55)26(51)20(14-47)62-38)23(43)6-10-44(17,16-43)67-39-35(65-37-32(57)29(54)25(50)19(13-46)61-37)34(27(52)21(15-48)63-39)64-36-31(56)28(53)24(49)18(12-45)60-36/h18-39,45-58H,1,4-16H2,2-3H3/t18-,19-,20-,21-,22+,23+,24-,25-,26-,27-,28+,29+,30+,31-,32-,33-,34+,35-,36+,37+,38+,39+,41-,42-,43-,44+/m1/s1
Formel vikt 966.4308
Procent renhet >95%