Learn More
Fisherbrand™ Recirculating Chiller for Sonicators Accessories: Tubing/Connector Set
Accessories for recirculating chiller
Brand: Fisherbrand™
Additional Details : Weight : 0.45000kg
Description
Tubing and connector set for use with Qsonica Recirculating Chiller for Sonicators.
Tubing/Connector Set for Cup Horn
- Quick-connect tubing and fittings (sold separately) are used to attach Chiller (Cat. No. 15-338-421) to ports on the cup horn
- Allows cold water to recirculate through the ultrasonic horn reservoir
Specifications
Tubing and Connector Set | |
Ideal for attach chiller to ports on the cup horn | |
Q500 and Q700 Sonicator™ | |
2 male fittings, 2 right angle male fittings and 2 female fittings |