missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant A. victoria Aequorea victoria GFP Protein
A DNA sequence encoding the Aequorea victoria (Jellyfish) Aequorea victoria GFP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7182-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
GFP Protein | |
Research Use Only | |
GFP | |
Recombinant Protein | |
E. coli | |
MSKGEELFTG VVPILVELDG DVNGHKFSVS GEGEGDATYG KLTLKFICTT GKLPVPWPTL VTTFSYGVQC FSRYPDHMKQ HDFFKSAMPE GYVQERTIFF KDDGNYKTRA EVKFEGDTLV NRIELKGIDF KEDGNILGHK LEYNYNSHNV YIMADKQKNG IKVNFKIRHN IEDGSVQLAD HYQQNTPIGD GPVLLPDNHY LSTQSALSKD PNEKRDHMVL LEFVTAAGIT HGMDELYK | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
A. victoria | |
His | |
Tris-based buffer, 50% glycerol |