missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant B. abortus vjbR Protein

A DNA sequence encoding the Brucella abortus (strain 2308) vjbR, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7642-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16019990

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_002966460.1
vjbR Protein
Research Use Only
vjbR
Recombinant Protein
Yeast
MSLDLVHFPN YKKTFFGSSF QSDTLALLTR IRDEIGCRYV THTYRGRVGD CTKVNSADLT VLMTLPATWV ARYSSKNYFA IDPVFQEDAP YYRNDTSAIA RDLKEDADIC PAVAELLHDA EKHGLGNLFI AVSARNPKGV AGCTVFTFEV EDEDRTQFLA RMRPRLLSLA GIIHGTVCGC KDANSVASLL TPREVDCLRW AANGKTDGEI AEILSIARWT VVTYLQNAKI KLNCSNRTSA VATALSLGII DMPEVQHLV
Greater than 90% as determined by SDS-PAGE.
29595784
200 μg
Not determined.
Full Length
B. abortus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers