missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Attachment protein G3P Protein
A DNA sequence encoding the Enterobacteria phage M13 (Bacteriophage M13) Attachment protein G3P, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9483-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_510891.1 | |
Attachment protein G3P Protein | |
Research Use Only | |
III | |
Recombinant Protein | |
Yeast | |
AETVESCLAK PHTENSFTNV WKDDKTLDRY ANYEGCLWNA TGVVVCTGDE TQCYGTWVPI GLAIPENEGG GSEGGGSEGG GSEGGGTKPP EYGDTPIPGY TYINPLDGTY PPGTEQNPAN PNPSLEESQP LNTFMFQNNR FRNRQGALTV YTGTVTQGTD PVKTYYQYTP VSSKAMYDAY WNGKFRDCAF HSGFNEDPFV CEYQGQSSDL PQPPVNAGGG SGGGSGGGSE GGGSEGGGSE GGGSEGGGSG GGSGSGDFDY EKMANANKGA MTENADENAL QSDAKGKLDS VATDYGAAID GFIGDVSGLA NGNGATGDFA GSNSQMAQVG DGDNSPLMNN FRQYLPSLPQ SVECRPFVFS AGKPYEFSID CDKINLFRGV FAFLLYVATF MYVFSTFANI LRNKES | |
Greater than 90% as determined by SDS-PAGE. |
927334 | |
200 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His | |
Tris-based buffer, 50% glycerol |