missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Recombination and repair Protein

A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Recombination and repair protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7355-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058400

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_049656.2
Recombination and repair Protein
Research Use Only
UVSX
Recombinant Protein
E. coli
MSDLKSRLIK ASTSKLTAEL TASKFFNEKD VVRTKIPMMN IALSGEITGG MQSGLLILAG PSKSFKSNFG LTMVSSYMRQ YPDAVCLFYD SEFGITPAYL RSMGVDPERV IHTPVQSLEQ LRIDMVNQLD AIERGEKVVV FIDSLGNLAS KKETEDALNE KVVSDMTRAK TMKSLFRIVT PYFSTKNIPC IAINHTYETQ EMFSKTVMGG GTGPMYSADT VFIIGKRQIK DGSDLQGYQF VLNVEKSRTV KEKSKFFIDV KFDGGIDPYS GLLDMALELG FVVKPKNGWY AREFLDEETG EMIREEKSWR AKDTNCTTFW GPLFKHQPFR DAIKRAYQLG AIDSNEIVEA EVDELINSKV EKFKSPESKS KSAADLETDL EQLSDMEEFN E
Greater than 86.2% as determined by SDS-PAGE.
1258704
10 μg
Not determined.
Full Length
Bacteriophage
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers