missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Adiponectin / Acrp30 / ADIPOQ Protein
A DNA sequence encoding the Bos taurus (Bovine) Adiponectin / Acrp30 / ADIPOQ, was expressed in the hosts and tags indicated.
3070.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_777167.1 |
---|---|
Gene ID (Entrez) | 282865 |
Name | Adiponectin / Acrp30 / ADIPOQ Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049990
|
enQuireBio™
QP7647-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069990
|
enQuireBio™
QP7647-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059990
|
enQuireBio™
QP7647-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079990
|
enQuireBio™
QP7647-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089990
|
enQuireBio™
QP7647-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099990
|
enQuireBio™
QP7647-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_777167.1 | |
Adiponectin / Acrp30 / ADIPOQ Protein | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
282865 | |
Research Use Only | |
ADIPOQ | |
Recombinant Protein | |
E. coli | |
EDNMEDPPLP KGACAGWMAG IPGHPGHNGT PGRDGRDGTP GEKGEKGDPG LVGPKGDTGE TGITGIEGPR GFPGTPGRKG EPGESAYVYR SAFSVGLERQ VTVPNVPIRF TKIFYNQQNH YDGTTGKFLC NIPGLYYFSY HITVYLKDVK VSLYKNDKAL LFTHDQFQDK NVDQASGSVL LYLEKGDQVW LQVYEGENHN GVYADNVNDS TFTGFLLYHN IVE | |
Greater than 90% as determined by SDS-PAGE. |